Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977694.1 | XP_027833495.1 | 8 - 9 | 900 | 106 | |
| XM_027977695.1 | XP_027833496.1 | 8 - 9 | 916 | 106 | ||
| XM_027977696.1 | XP_027833497.1 | 8 - 9 | 929 | 106 | ||
More Information
Set Membership: |
|

CIBAR2
Genomic Map