Assay Details
| Gene Symbol: ECHDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099541.2 | XP_014955027.2 | 1 - 2 | 366 | 65 | |
| XM_027976531.1 | XP_027832332.1 | 1 - 2 | 366 | 65 | ||
More Information
Set Membership: |
|

ECHDC3
Genomic Map