Assay Details
| Gene Symbol: PLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957472.1 | XP_027813273.1 | 8 - 9 | 1344 | 69 | |
| XM_027957473.1 | XP_027813274.1 | 8 - 9 | 1338 | 69 | ||
More Information
Set Membership: |
|

PLD4
Genomic Map