Assay Details
| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973415.1 | XP_027829216.1 | 2 - 3 | 1699 | 97 | |
| XM_027973416.1 | XP_027829217.1 | 3 - 4 | 1612 | 97 | ||
| XM_027973417.1 | XP_027829218.1 | 3 - 4 | 1591 | 97 | ||
| XM_027973418.1 | XP_027829219.1 | 4 - 5 | 1546 | 97 | ||
| XM_027973419.1 | XP_027829220.1 | 4 - 5 | 1525 | 97 | ||
| XM_027973420.1 | XP_027829221.1 | 3 - 4 | 1421 | 97 | ||
More Information
Set Membership: |
|

MFSD3
Genomic Map