Assay Details
Gene Symbol: SNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977734.1 | XP_027833535.1 | 3 | 2105 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
