Assay Details
Gene Symbol: NLRP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978000.1 | XP_027833801.1 | 9 - 10 | 3038 | 79 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NLRP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978000.1 | XP_027833801.1 | 9 - 10 | 3038 | 79 |