Assay Details
| Gene Symbol: PAIP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006056.4 | XP_004006105.1 | 3 - 4 | 506 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAIP2B| Gene Symbol: PAIP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006056.4 | XP_004006105.1 | 3 - 4 | 506 | 62 | |
Set Membership: |
|