Assay Details
| Gene Symbol: SLC35A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011288.4 | XP_004011337.1 | 4 - 5 | 691 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC35A1| Gene Symbol: SLC35A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011288.4 | XP_004011337.1 | 4 - 5 | 691 | 83 | |
Set Membership: |
|