Assay Details
| Gene Symbol: SLC22A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011439.4 | XP_004011488.2 | 7 - 8 | 1617 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A2| Gene Symbol: SLC22A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011439.4 | XP_004011488.2 | 7 - 8 | 1617 | 63 | |
Set Membership: |
|