Assay Details
| Gene Symbol: APOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015314.4 | XP_004015363.2 | 3 - 4 | 501 | 53 | |
| XM_027978462.1 | XP_027834263.1 | 4 - 5 | 2577 | 53 | ||
| Gene Symbol: APOC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015313.4 | XP_004015362.2 | 5 - 6 | 1360 | 53 | |
| XM_027978463.1 | XP_027834264.1 | 4 - 5 | 2604 | 53 | ||
More Information
Set Membership: |
|

APOC2
Genomic Map