Assay Details
| Gene Symbol: NECTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978456.1 | XP_027834257.1 | 4 - 5 | 1541 | 74 | |
| XM_027978457.1 | XP_027834258.1 | 4 - 5 | 1542 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NECTIN2| Gene Symbol: NECTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978456.1 | XP_027834257.1 | 4 - 5 | 1541 | 74 | |
| XM_027978457.1 | XP_027834258.1 | 4 - 5 | 1542 | 74 | ||