Assay Details
| Gene Symbol: LOC101117536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980289.1 | XP_027836090.1 | 2 - 3 | 206 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117536| Gene Symbol: LOC101117536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980289.1 | XP_027836090.1 | 2 - 3 | 206 | 60 | |
Set Membership: |
|