Assay Details
| Gene Symbol: ARHGAP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021555.4 | XP_004021604.2 | 8 - 9 | 1192 | 59 | |
| XM_012105509.3 | XP_011960899.2 | 8 - 9 | 1240 | 59 | ||
| XM_012105513.2 | XP_011960903.2 | 6 - 7 | 1164 | 59 | ||
More Information
Set Membership: |
|

ARHGAP22
Genomic Map