Assay Details
| Gene Symbol: FUT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021766.4 | XP_004021815.2 | 4 - 5 | 1640 | 66 | |
| XM_027962714.1 | XP_027818515.1 | 3 - 4 | 1285 | 66 | ||
More Information
Set Membership: |
|

FUT10
Genomic Map