Assay Details
| Gene Symbol: CNTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958158.1 | XP_027813959.1 | 11 - 12 | 1509 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNTN4| Gene Symbol: CNTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958158.1 | XP_027813959.1 | 11 - 12 | 1509 | 65 | |
Set Membership: |
|