Assay Details
| Gene Symbol: ALG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968233.1 | XP_027824034.1 | 11 - 12 | 1691 | 63 | |
| XM_027968235.1 | XP_027824036.1 | 11 - 12 | 1667 | 63 | ||
| XM_027968236.1 | XP_027824037.1 | 11 - 12 | 1628 | 63 | ||
| XM_027968237.1 | XP_027824038.1 | 8 - 9 | 2617 | 63 | ||
| XM_027968238.1 | XP_027824039.1 | 9 - 10 | 1535 | 63 | ||
| XM_027968239.1 | XP_027824040.1 | 9 - 10 | 1455 | 63 | ||
More Information
Set Membership: |
|

ALG12
Genomic Map