Assay Details
| Gene Symbol: LOC101105047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103812.2 | XP_014959298.2 | 4 - 5 | 1272 | 95 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101105047| Gene Symbol: LOC101105047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103812.2 | XP_014959298.2 | 4 - 5 | 1272 | 95 | |