Assay Details
| Gene Symbol: LOC101108282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962515.1 | XP_027818316.1 | 18 - 19 | 3272 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101108282| Gene Symbol: LOC101108282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962515.1 | XP_027818316.1 | 18 - 19 | 3272 | 82 | |