Assay Details
| Gene Symbol: LOC114115563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971515.1 | XP_027827316.1 | 7 - 8 | 1467 | 114 | |
| Gene Symbol: LOC101111971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963821.1 | XP_027819622.1 | 12 - 13 | 2264 | 114 | |
More Information
Set Membership: |
|

LOC114115563
Genomic Map