Assay Details
| Gene Symbol: WHAMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101910.2 | XP_014957396.2 | 5 - 6 | 1363 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WHAMM| Gene Symbol: WHAMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101910.2 | XP_014957396.2 | 5 - 6 | 1363 | 72 | |
Set Membership: |
|