Assay Details
| Gene Symbol: LOC101112575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973279.1 | XP_027829080.1 | 10 - 11 | 806 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101112575| Gene Symbol: LOC101112575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973279.1 | XP_027829080.1 | 10 - 11 | 806 | 81 | |