Assay Details
| Gene Symbol: CMTM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957943.1 | XP_027813744.1 | 3 - 4 | 466 | 80 | |
| XM_027957944.1 | XP_027813745.1 | 3 - 4 | 352 | 80 | ||
More Information
Set Membership: |
|

CMTM8
Genomic Map