Assay Details
| Gene Symbol: CIBAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012184137.3 | XP_012039527.2 | 4 - 5 | 577 | 143 | |
| XM_012184138.3 | XP_012039528.2 | 4 - 5 | 577 | 143 | ||
More Information
Set Membership: |
|

CIBAR1
Genomic Map