Assay Details
| Gene Symbol: ASXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099825.2 | XP_014955311.2 | 5 - 6 | 415 | 74 | |
| XM_027977065.1 | XP_027832866.1 | 5 - 6 | 415 | 74 | ||
| XM_027977066.1 | XP_027832867.1 | 4 - 5 | 412 | 74 | ||
| XM_027977067.1 | XP_027832868.1 | 4 - 5 | 412 | 74 | ||
| XM_027977068.1 | XP_027832869.1 | 5 - 6 | 2600 | 74 | ||
| XM_027977069.1 | XP_027832870.1 | 5 - 6 | 415 | 74 | ||
More Information
Set Membership: |
|

ASXL1
Genomic Map