Assay Details
| Gene Symbol: CA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004279.4 | XP_004004328.1 | 5 - 6 | 2569 | 60 | |
| XM_012121475.3 | XP_011976865.1 | 5 - 6 | 2569 | 60 | ||
More Information
Set Membership: |
|

CA9
Genomic Map