Assay Details
| Gene Symbol: ATF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006355.3 | XP_004006404.1 | 5 - 6 | 804 | 66 | |
| XM_012174289.2 | XP_012029679.2 | 6 - 7 | 796 | 66 | ||
| XM_012174295.3 | XP_012029685.2 | 5 - 6 | 910 | 66 | ||
| XM_027967333.1 | XP_027823134.1 | 5 - 6 | 699 | 66 | ||
More Information
Set Membership: |
|

ATF1
Genomic Map