Assay Details
| Gene Symbol: IMMP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016367.4 | XP_004016416.1 | 6 - 7 | 675 | 75 | |
| XM_012096156.3 | XP_011951546.1 | 7 - 8 | 727 | 75 | ||
| XM_012096158.3 | XP_011951548.1 | 8 - 9 | 987 | 75 | ||
| XM_012096159.3 | XP_011951549.1 | 7 - 8 | 719 | 75 | ||
More Information
Set Membership: |
|

IMMP1L
Genomic Map