Assay Details
| Gene Symbol: FBXL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960479.1 | XP_027816280.1 | 2 - 3 | 363 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXL15| Gene Symbol: FBXL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960479.1 | XP_027816280.1 | 2 - 3 | 363 | 102 | |
Set Membership: |
|