Assay Details
| Gene Symbol: PTEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102490.3 | XP_011957880.1 | 4 - 5 | 1138 | 84 | |
| XM_027960248.1 | XP_027816049.1 | 8 - 9 | 1852 | 84 | ||
More Information
Set Membership: |
|

PTEN
Genomic Map