Assay Details
| Gene Symbol: LOC101121595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968820.1 | XP_027824621.1 | 5 - 6 | 1600 | 156 | |
| XM_027968821.1 | XP_027824622.1 | 5 - 6 | 1610 | 156 | ||
| XM_027968823.1 | XP_027824624.1 | 5 - 6 | 673 | 156 | ||
| XM_027968825.1 | XP_027824626.1 | 5 - 6 | 714 | 156 | ||
| XM_027968826.1 | XP_027824627.1 | 5 - 6 | 1597 | 156 | ||
| XM_027968827.1 | XP_027824628.1 | 5 - 6 | 701 | 156 | ||
| XM_027968828.1 | XP_027824629.1 | 5 - 6 | 693 | 156 | ||
More Information
Set Membership: |
|

LOC101121595
Genomic Map