Assay Details
| Gene Symbol: LOC101121595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968825.1 | XP_027824626.1 | 8 - 9 | 915 | 104 | |
| XM_027968826.1 | XP_027824627.1 | 8 - 9 | 1798 | 104 | ||
| XM_027968827.1 | XP_027824628.1 | 8 - 9 | 902 | 104 | ||
More Information
Set Membership: |
|

LOC101121595
Genomic Map