Assay Details
| Gene Symbol: DMWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978496.1 | XP_027834297.1 | 2 - 3 | 638 | 71 | |
| XM_027978497.1 | XP_027834298.1 | 2 - 3 | 638 | 71 | ||
More Information
Set Membership: |
|

DMWD
Genomic Map