Assay Details
Gene Symbol: LOC114110420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960726.1 | XP_027816527.1 | 2 - 3 | 226 | 62 |
Gene Symbol: LOC101120466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964020.1 | XP_027819821.1 | 19 - 20 | 2447 | 62 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
