Assay Details
| Gene Symbol: TRIM25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098442.2 | XP_014953928.2 | 5 - 6 | 1227 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM25| Gene Symbol: TRIM25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098442.2 | XP_014953928.2 | 5 - 6 | 1227 | 56 | |
Set Membership: |
|