Assay Details
| Gene Symbol: LOC101113696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977945.1 | XP_027833746.1 | 20 - 21 | 3281 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101113696| Gene Symbol: LOC101113696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977945.1 | XP_027833746.1 | 20 - 21 | 3281 | 56 | |
Set Membership: |
|