Assay Details
| Gene Symbol: ATP6AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962901.1 | XP_027818702.1 | 9 - 10 | 1210 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6AP1| Gene Symbol: ATP6AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962901.1 | XP_027818702.1 | 9 - 10 | 1210 | 62 | |
Set Membership: |
|