Assay Details
Gene Symbol: C4H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007949.3 | XP_004007998.2 | 2 - 3 | 256 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C4H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007949.3 | XP_004007998.2 | 2 - 3 | 256 | 67 |
Set Membership: |
![]() ![]() ![]() |