Assay Details
| Gene Symbol: TMEM43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957824.1 | XP_027813625.1 | 12 - 13 | 1388 | 64 | |
| XM_027957825.1 | XP_027813626.1 | 11 - 12 | 1183 | 64 | ||
More Information
Set Membership: |
|

TMEM43
Genomic Map