Assay Details
| Gene Symbol: LOC101118592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107055.2 | XP_011962445.1 | 4 - 5 | 354 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118592| Gene Symbol: LOC101118592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107055.2 | XP_011962445.1 | 4 - 5 | 354 | 68 | |
Set Membership: |
|