Assay Details
| Gene Symbol: TCFL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012188858.3 | XP_012044248.2 | 4 - 5 | 1255 | 57 | |
| XM_027976968.1 | XP_027832769.1 | 4 - 5 | 1252 | 57 | ||
More Information
Set Membership: |
|

TCFL5
Genomic Map