Assay Details
| Gene Symbol: NUMBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978301.1 | XP_027834102.1 | 4 - 5 | 515 | 60 | |
| XM_027978302.1 | XP_027834103.1 | 4 - 5 | 515 | 60 | ||
| XM_027978303.1 | XP_027834104.1 | 4 - 5 | 530 | 60 | ||
| XM_027978304.1 | XP_027834105.1 | 3 - 4 | 1351 | 60 | ||
More Information
Set Membership: |
|

NUMBL
Genomic Map