Assay Details
| Gene Symbol: GOLT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186925.3 | XP_012042315.2 | 4 - 5 | 467 | 87 | |
| XM_012186926.3 | XP_012042316.2 | 4 - 5 | 400 | 87 | ||
More Information
Set Membership: |
|

GOLT1A
Genomic Map