Assay Details
Gene Symbol: SOX13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976187.1 | XP_027831988.1 | 13 - 14 | 2450 | 74 | |
XM_027976188.1 | XP_027831989.1 | 13 - 14 | 2447 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
