Assay Details
| Gene Symbol: FAM234B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967852.1 | XP_027823653.1 | 11 - 12 | 1698 | 72 | |
| XM_027967853.1 | XP_027823654.1 | 11 - 12 | 1695 | 72 | ||
More Information
Set Membership: |
|

FAM234B
Genomic Map