Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097311.2 | XP_014952797.2 | 23 - 24 | 5642 | 96 | |
| XM_027972311.1 | XP_027828112.1 | 23 - 24 | 5642 | 96 | ||
| XM_027972312.1 | XP_027828113.1 | 22 - 23 | 5612 | 96 | ||

DOP1A
Genomic Map