Assay Details
| Gene Symbol: TBXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011450.4 | XP_004011499.1 | 4 - 5 | 668 | 61 | |
| XM_027972732.1 | XP_027828533.1 | 4 - 5 | 668 | 61 | ||
| XM_027972733.1 | XP_027828534.1 | 4 - 5 | 668 | 61 | ||
More Information
Set Membership: |
|

TBXT
Genomic Map