Assay Details
| Gene Symbol: SLC39A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974012.1 | XP_027829813.1 | 4 - 5 | 413 | 53 | |
| XM_027974013.1 | XP_027829814.1 | 4 - 5 | 413 | 53 | ||
More Information
Set Membership: |
|

SLC39A11
Genomic Map