Assay Details
| Gene Symbol: RSKR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013227.4 | XP_004013276.1 | 7 - 8 | 972 | 71 | |
| XM_012185397.3 | XP_012040787.1 | 7 - 8 | 1026 | 71 | ||
| XM_015098538.2 | XP_014954024.1 | 6 - 7 | 921 | 71 | ||
| XM_015098539.2 | XP_014954025.1 | 6 - 7 | 867 | 71 | ||
| XM_027975232.1 | XP_027831033.1 | 7 - 8 | 1044 | 71 | ||
| XM_027975233.1 | XP_027831034.1 | 7 - 8 | 990 | 71 | ||
More Information
Set Membership: |
|

RSKR
Genomic Map