Assay Details
| Gene Symbol: TMEM241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020478.4 | XP_004020527.2 | 3 - 4 | 254 | 57 | |
| XM_012103488.3 | XP_011958878.2 | 3 - 4 | 254 | 57 | ||
| XM_012103490.3 | XP_011958880.1 | 3 - 4 | 254 | 57 | ||
More Information
Set Membership: |
|

TMEM241
Genomic Map