Assay Details
| Gene Symbol: CYRIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012164859.3 | XP_012020249.1 | 6 - 7 | 1113 | 83 | |
| XM_015094234.2 | XP_014949720.1 | 6 - 7 | 654 | 83 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYRIA| Gene Symbol: CYRIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012164859.3 | XP_012020249.1 | 6 - 7 | 1113 | 83 | |
| XM_015094234.2 | XP_014949720.1 | 6 - 7 | 654 | 83 | ||